missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM46A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55488
This item is not returnable.
View return policy
Description
FAM46A Polyclonal specifically detects FAM46A in Human samples. It is validated for Western Blot.
Specifications
| FAM46A | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| C6orf37, chromosome 6 open reading frame 37, family with sequence similarity 46, member A, FLJ20037, FLJ31495, HBV XAg-transactivated protein 11, HBV X-transactivated gene 11 protein, hypothetical protein LOC55603, retinal expressed gene C6orf37, XTP11 | |
| Rabbit | |
| 50 kDa | |
| 100 μL | |
| Primary | |
| Zebrafish 83%. | |
| Human, Mouse, Rat, Pig, Equine, Yeast, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96IP4 | |
| TENT5A | |
| Synthetic peptides corresponding to FAM46A(family with sequence similarity 46, member A) The peptide sequence was selected from the N terminal of FAM46A. Peptide sequence MAEGEGYFAMSEDELACSPYIPLGGDFGGGDFGGGDFGGGDFGGGGSFGG. | |
| Affinity purified | |
| RUO | |
| 55603 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction