Learn More
Falcon™ Pipet Controller Hydrophobic PTFE Filters
Replacement hydrophobic PTFE disc filters
Brand:  Falcon™ 357473
Code : 67
Additional Details : Weight : 0.01800kg
 View more versions of this product
Product Code. 15339840
  • £44.49 / Pack of 5
Estimated Shipment: 19-09-2024
to see stock.
Add to basket
Description
Description
Additional filters are available as standard accessories. The filters, pipet holder, and nose piece are autoclavable.
Specifications
Specifications
PTFE
0.45 μm
For use with pipet controller
5/Cs.