missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Falcon™ Pipet Controller Hydrophobic PTFE Filters
Replacement hydrophobic PTFE disc filters
£44.49
Specifications
Material | PTFE |
---|---|
For Use With (Equipment) | For use with pipet controller |
Quantity | 5/Cs. |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
15339840
|
Falcon™
357473 |
0.45 μm |
£44.49
Pack of 5 |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||||
15329840
|
Falcon™
357472 |
0.2 μm |
£44.49
Pack of 5 |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||||
Description
Additional filters are available as standard accessories. The filters, pipet holder, and nose piece are autoclavable.
Specifications
PTFE | |
For use with pipet controller | |
5/Cs. |