missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAF2 Mouse anti-Human, Clone: 3H6D2, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | FAF2 |
|---|---|
| Clone | 3H6D2 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16848685
|
Proteintech
66629-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16838685
|
Proteintech
66629-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is highly expressed in peripheral blood of patients with atopic dermatitis (AD), compared to normal individuals. It may play a role in regulating the resistance to apoptosis that is observed in T cells and eosinophils of AD patients.Specifications
| FAF2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| ETEA, FAF2, FAS associated factor 2, KIAA0887, Protein ETEA, UBXD8, UBXN3B | |
| FAF2 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 3H6D2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q96CS3 | |
| 23197 | |
| FAF2 Fusion Protein Ag21536 | |
| Primary | |
| -20°C | |
| FAF2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title