missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAF2 Mouse anti-Human, Clone: 3H6D2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66629-1-IG-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is highly expressed in peripheral blood of patients with atopic dermatitis (AD), compared to normal individuals. It may play a role in regulating the resistance to apoptosis that is observed in T cells and eosinophils of AD patients.Specifications
| FAF2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q96CS3 | |
| FAF2 | |
| FAF2 Fusion Protein Ag21536 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 3H6D2 | |
| Unconjugated | |
| FAF2 | |
| ETEA, FAF2, FAS associated factor 2, KIAA0887, Protein ETEA, UBXD8, UBXN3B | |
| Mouse | |
| Protein G | |
| RUO | |
| 23197 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction