missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor IX Antibody (3G3), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Factor IX |
|---|---|
| Clone | 3G3 |
| Dilution | Western Blot 1:500-1:5000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043388
|
Novus Biologicals
NBP3-26593-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043883
|
Novus Biologicals
NBP3-26593-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Factor IX Monoclonal antibody specifically detects Factor IX in Human samples. It is validated for Western Blot, ELISASpecifications
| Factor IX | |
| Western Blot 1:500-1:5000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2158 | |
| IgG | |
| Affinity purified |
| 3G3 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Christmas factor, coagulation factor IX, EC 3.4.21, EC 3.4.21.22, factor 9, FIX, HEMB, MGC129641, MGC129642, plasma thromboplastic component, Plasma thromboplastin component, PTC | |
| A synthesized peptide derived from Human Factor IX [UniProt P00740] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title