Learn More
Factor IX Antibody (3G3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26593-100ul
 View more versions of this product
Product Code. 30043883
  • £406.00 / 100µL
Description
Factor IX Monoclonal antibody specifically detects Factor IX in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Factor IX
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
2158
Store at -20 to -70C. Avoid freeze-thaw cycles.
IgG
Western Blot, ELISA
3G3
Western Blot 1:500-1:5000, ELISA
Christmas factor, coagulation factor IX, EC 3.4.21, EC 3.4.21.22, factor 9, FIX, HEMB, MGC129641, MGC129642, plasma thromboplastic component, Plasma thromboplastin component, PTC
A synthesized peptide derived from Human Factor IX [UniProt P00740]
100 μL
Primary
Human
Purified