missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FACA/FANCA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£164.00 - £288.00
Specifications
| Antigen | FACA/FANCA |
|---|---|
| Dilution | Western Blot 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18044685
|
Novus Biologicals
NB100-2564SS |
0.025 mL |
£164.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18067563
|
Novus Biologicals
NB100-2564 |
0.1 mL |
£288.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FACA/FANCA Polyclonal specifically detects FACA/FANCA in Human samples. It is validated for Western Blot.Specifications
| FACA/FANCA | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
| Whole antisera with No Preservative | |
| FA, FA1, FAAMGC75158, FACAFanconi anemia group A protein, FAH, FA-H, FANCH, Fanconi anemia, complementation group A, Fanconi anemia, complementation group H, Fanconi anemia, type 1, Protein FACA | |
| FANCA | |
| IgG | |
| Unpurified | |
| 170 kDa |
| Western Blot 1:1000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| O15360 | |
| 2175 | |
| A synthetic peptide within residues 1200-1300 of the human FANCA protein. [Swiss-Prot# O15360] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title