missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FACA/FANCA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-2564
This item is not returnable.
View return policy
Description
FACA/FANCA Polyclonal specifically detects FACA/FANCA in Human samples. It is validated for Western Blot.
Specifications
| FACA/FANCA | |
| Polyclonal | |
| Western Blot 1:1000 | |
| O15360 | |
| FANCA | |
| A synthetic peptide within residues 1200-1300 of the human FANCA protein. [Swiss-Prot# O15360] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Antisera |
| Western Blot | |
| Unconjugated | |
| Whole antisera with No Preservative | |
| FA, FA1, FAAMGC75158, FACAFanconi anemia group A protein, FAH, FA-H, FANCH, Fanconi anemia, complementation group A, Fanconi anemia, complementation group H, Fanconi anemia, type 1, Protein FACA | |
| Rabbit | |
| 170 kDa | |
| 0.1 mL | |
| Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
| 2175 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction