missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP2/I-FABP Antibody (FABP2/7670), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20497-20ug
This item is not returnable.
View return policy
Description
FABP2/I-FABP Monoclonal antibody specifically detects FABP2/I-FABP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FABP2/I-FABP | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein | |
| Recombinant full length human FABP2/I-FABP protein | |
| 20 μg | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| 2169 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| FABP2/7670 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction