missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP2/I-FABP Antibody (FABP2/7670), Novus Biologicals™
Mouse Monoclonal Antibody
£195.00 - £385.00
Specifications
| Antigen | FABP2/I-FABP |
|---|---|
| Clone | FABP2/7670 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620044
|
Novus Biologicals
NBP3-20497-20ug |
20 μg |
£195.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18670275
|
Novus Biologicals
NBP3-20497-100ug |
100 μg |
£385.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FABP2/I-FABP Monoclonal antibody specifically detects FABP2/I-FABP in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| FABP2/I-FABP | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| 10mM PBS with 0.05% BSA | |
| 2169 | |
| IgG | |
| Protein A or G purified |
| FABP2/7670 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, I-FABPMGC133132, Intestinal-type fatty acid-binding protein | |
| Recombinant full length human FABP2/I-FABP protein | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title