Description
- The FABian™ cell selection system is a fully automatic bench top instrument, which utilizes IBA’s reversible Fab column-based Traceless Affinity Cell Selection (Fab-TACS™) procedure for non-magnetic isolation of PBMCs, T, B and other cells of interest with high efficiency and purity directly from whole blood, buffy coat or other single-cell tissue suspensions.
Specifications
Specifications
| Includes | Automated cell selection system with 10 mL syringe adapter |
| For Use With (Application) | PBMC, T and B Cell Isolation |
| Quantity | 1 |