missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ezrin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-52977SS
This item is not returnable.
View return policy
Description
Ezrin Polyclonal antibody specifically detects Ezrin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Ezrin | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2MGC1584, villin 2 (ezrin), villin-2 | |
| Partial recombinant human Ezrin protein (amino acids 438-562) [UniProt P15311] | |
| 0.025 mg | |
| Cancer, Cell Cycle and Replication, Signal Transduction | |
| 7430 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1.0 μg/mL, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/ Immunofluorescence 5 μg/mL, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
| P15311 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction