missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ezrin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £395.00
Specifications
| Antigen | Ezrin |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1.0 μg/mL, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/ Immunofluorescence 5 μg/mL, Immunohistochemistry-Paraffin 1:100 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18653119
|
Novus Biologicals
NBP2-52977SS |
0.025 mg |
£169.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18650479
|
Novus Biologicals
NBP2-52977 |
0.1 mg |
£395.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ezrin Polyclonal antibody specifically detects Ezrin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Ezrin | |
| Western Blot 1.0 μg/mL, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/ Immunofluorescence 5 μg/mL, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| P15311 | |
| 7430 | |
| IgG | |
| Immunogen affinity purified |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Signal Transduction | |
| PBS | |
| CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2MGC1584, villin 2 (ezrin), villin-2 | |
| Partial recombinant human Ezrin protein (amino acids 438-562) [UniProt P15311] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title