Learn More
Biochrom EZ Read 800 Microplate Reader
Description
The EZ Read 800 is a fast and accurate microplate reader that is an ideal choice to keep up with a heavy workload in a busy high-throughput laboratory.
The EZ Read 800 microplate readers are PC-controlled using new, intuitive Galapagos software.
EZ Read 800 ELISA and EZ Read 800 Research microplate reader features:
- >5 second for 96-well plate
- Measures flat, u-bottom and v-bottom plate with standard ANSI/SBS footprint
- Four modes of shaking for complete mixing of solutions and more reproducible results
- Automatic lamp calibration ensures measurement performance
- USB 2.0 PC connection
EZ Read 800 comes in two applications focused models:
- Biochrom EZ Read 800 ELISA: a dedicated ELISA reader. Comes with 4 filters that can be used to measure most ELISA assays (405, 450, 492 and 620 nm)
- ELISA Assay Substrate
- NPP, ABTS use Filter 405 nm
- OPD use Filter 492 nm
- Slow TMB, Turbo TMB, Ultra TMB use Filter 450 nm
- Reference wavelength use Filter 620 nm
- ELISA Assay Substrate
- Biochrom EZ Read 800 Research: offers versatility by extending its applications capacity to measuring total protein and cell proliferation assays in additional to ELISA assays. Comes with 6 filters that can be used to measure ELISA, total protein and cell proliferation assays (405, 450, 492, 562, 595 and 620 nm).
- ELISA Assay Substrate
- NPP, ABTS use Filter 405 nm
- OPD use Filter 492 nm
- Slow TMB, Turbo TMB, Ultra TMB use Filter 450 nm
- Reference wavelength use Filter 620 nm
- Total Protein Assays
- Bradford use Filter 595 nm
- BCA use Filter 562 nm
- Cell Proliferation/Viability Assays
- XTT use Filter 492 nm
- Cell Density use Filter 620 nm
- ELISA Assay Substrate
Galapagos for EZ Read Software's 'click and drag' approach makes usability a key feature.
- Create new plate layouts
- Add multiple assays to a plate
- Add multiple plates per analysis
- Easy to create new methods
- Select only the data you want for data reporting
- Windows 7 & 8 compatibility as well as older Windows OS from XP
- So easy you won't need to export your data into Excel
Specifications
Specifications
| Detector Type | 8 Silicon Photodiodes |
| Light Source | Tungsten Halogen Lamp |
| Measurement Speed | <5 sec. |
| Plate Type | 96-well Plates |
| Shaking | 4 speed standard |
| Software Type | Galapagos Software |
| Wavelength Selection | Filters |
| Accuracy | 1% from 0.100 to 2.5 OD |
| Electrical Requirements | 90/250 V, 50/60 Hz, 80 VA |
| Filters | 405, 450, 492 and 620 nm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
