missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biochrom EZ Read 800 Microplate Reader
Click to view available options
Model:
EZ Read 800 ELISA
EZ Read 800 RESEARCH
Unit Size:
Each
Description
The EZ Read 800 is a fast and accurate microplate reader that is an ideal choice to keep up with a heavy workload in a busy high-throughput laboratory.
The EZ Read 800 microplate readers are PC-controlled using new, intuitive Galapagos software.
EZ Read 800 ELISA and EZ Read 800 Research microplate reader features:
- >5 second for 96-well plate
- Measures flat, u-bottom and v-bottom plate with standard ANSI/SBS footprint
- Four modes of shaking for complete mixing of solutions and more reproducible results
- Automatic lamp calibration ensures measurement performance
- USB 2.0 PC connection
EZ Read 800 comes in two applications focused models:
- Biochrom EZ Read 800 ELISA: a dedicated ELISA reader. Comes with 4 filters that can be used to measure most ELISA assays (405, 450, 492 and 620 nm)
- ELISA Assay Substrate
- NPP, ABTS use Filter 405 nm
- OPD use Filter 492 nm
- Slow TMB, Turbo TMB, Ultra TMB use Filter 450 nm
- Reference wavelength use Filter 620 nm
- ELISA Assay Substrate
- Biochrom EZ Read 800 Research: offers versatility by extending its applications capacity to measuring total protein and cell proliferation assays in additional to ELISA assays. Comes with 6 filters that can be used to measure ELISA, total protein and cell proliferation assays (405, 450, 492, 562, 595 and 620 nm).
- ELISA Assay Substrate
- NPP, ABTS use Filter 405 nm
- OPD use Filter 492 nm
- Slow TMB, Turbo TMB, Ultra TMB use Filter 450 nm
- Reference wavelength use Filter 620 nm
- Total Protein Assays
- Bradford use Filter 595 nm
- BCA use Filter 562 nm
- Cell Proliferation/Viability Assays
- XTT use Filter 492 nm
- Cell Density use Filter 620 nm
- ELISA Assay Substrate
Galapagos for EZ Read Software's 'click and drag' approach makes usability a key feature.
- Create new plate layouts
- Add multiple assays to a plate
- Add multiple plates per analysis
- Easy to create new methods
- Select only the data you want for data reporting
- Windows 7 & 8 compatibility as well as older Windows OS from XP
- So easy you won't need to export your data into Excel
Specifications
Specifications
| Detector Type | 8 Silicon Photodiodes |
| Light Source | Tungsten Halogen Lamp |
| Measurement Speed | <5 sec. |
| Plate Type | 96-well Plates |
| Shaking | 4 speed standard |
| Software Type | Galapagos Software |
| Wavelength Selection | Filters |
| Accuracy | 1% from 0.100 to 2.5 OD |
| Electrical Requirements | 90/250 V, 50/60 Hz, 80 VA |
| Filters | 405, 450, 492, 562, 595 and 620 nm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
