missing translation for 'onlineSavingsMsg'
Learn More

Biochrom EZ Read 400 Microplate Reader

Product Code. 12664795
Change view
Click to view available options
Model:
EZ Read 400 ELISA
EZ Read 400 RESEARCH
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
12664795 EZ Read 400 RESEARCH Each
12694795 EZ Read 400 ELISA Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12664795 Supplier Biochrom Supplier No. 80400141

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

EZ Read 400 Microplate Readers with Galapagos for EZ Read Software.

The EZ Read 400 is a simple and robust microplate reader that is an ideal choice for laboratories which require a workhorse instrument for their routine absorbance assays. 

The EZ Read 400 microplate readers are PC-controlled using new, intuitive Galapagos software created by Biochrom to simplify data acquistion and analysis.

EZ Read 400 ELISA and EZ Read 800 Research microplate reader features

  • Measures flat, round and v-bottomed 96-well microplates and stripwells with standard ANSI SBS footprints are measured
  • Accurate measurements ensured by automatic lamp calibration prior to each reading
  • USB 2.0 PC connection
  • Interference filters for wavelength selection
  • Visible range (400 to 750 nm)

The EZ Read 400 is available in two applictions-focussed models:

  • EZ Read 400 ELISA is a dedicated ELISA reader. Comes with 4 filters that can be used to measure most ELISA assays (405, 450, 492 and 620 nm)
    • ELISA Assay Substrate
      • NPP, ABTS use Filter 405 nm
      • OPD use Filter 492 nm
      • Slow TMB, Turbo TMB, Ultra TMB use Filter 450 nm
      • Reference wavelength use Filter 620 nm
  • EZ Read 400 Research offers versatility by extending its applications capacity to measuring total protein, cell proliferation assays as well ELISA assays. Comes with 6 filters that can be used to measure ELISA, total protein and cell proliferation assays (405, 450, 492, 562, 570, 595, 620 and 650 nm)
    • ELISA Assay Substrate
      • NPP, ABTS use Filter 405 nm
      • OPD use Filter 492 nm
      • Slow TMB, Turbo TMB, Ultra TMB use Filter 450 nm
      • Reference wavelength use Filter 620 nm
    • Total Protein Assays
      • Bradford use Filter 595 nm
      • BCA use Filter 562 nm
      • Lowry use Filter 450 nm
      • Reference wavelength use Filter 650 nm
    • Cell Proliferation/Viability Assays
      • MTT use Filter 570 nm
      • XTT use Filter 492 nm
      • Cell Density use Filter 620 nm

Galapagos for EZ Read Software's 'click and drag' approach makes usability a key feature:

  • Create new plate layouts
  • Add multiple assays to a plate
  • Add multiple plates per analysis
  • Easy to create new methods
  • Select only the data you want for data reporting
  • Windows 7 & 8 compatibility as well as older Windows OS from XP
  • So easy you won't need to export your data into Excel

Specifications

Product Type Microplate Reader
Detector Type Single Channel Silicon Photodiode
Dimensions (L x W x H) 18 x 31.5 x 43.5 cm
Light Source Tungsten Halogen Lamp
Measurement Speed 25 sec.
Measurement Technology Absorbance
Plate Type 96-well Plates
Software Type Galapagos Software
Wavelength Selection Filters
Accuracy 0.5% at 1.0 OD (at 450 nm)
Description EZ Read 400 Research Microplate Reader
Electrical Requirements 100/240 V, 50/60 Hz, 1.5 A
Filters 405, 450, 492, 620 nm
For Use With (Application) ELISA
Linearity 0.25% and 0.0025 O.D. from 0.1 to 2.5 O.D. (at 492 nm)
Model EZ Read 400 RESEARCH
Precision 0.25% at 1.0 O.D. (at 450 nm)
Warranty 1 Year
Wavelength Range 400 to 750 nm
Weight (English) 14.5 lb.
Weight (Metric) 6.6 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.