missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Sulfo-NHS-SS-Biotin, No Weigh Format

Product Code. 10361554 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
8 x 1 mg
100 mg
10 x 1 mg
Unit Size:
100mg
10mg
Pack of 8
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10361554 8 x 1 mg Pack of 8
15925521 10 x 1 mg 10mg
11811205 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10361554 Supplier Thermo Scientific™ Supplier No. 21328

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermo Scientific™ EZ-Link™ Sulfo-NHS-SS-Biotin No-Weigh™ Format is a water-soluble, NHS-ester biotinylation reagent whose spacer arm includes a cleavable disulfide bond for reversible labeling of proteins and cell surface primary amines.

Thermo Scientific™ EZ-Link Sulfo-NHS-SS-Biotin enables simple and efficient biotin labeling of antibodies, proteins and any other primary amine-containing biomolecules in solution.

Highlights:

  • Cleavable analog of Sulfo-NHS-LC-Biotin
  • Can remove biotin label by cleaving spacer disulfide bond with reducing agents, such as DTT
  • Ideal for affinity purification experiments that require efficient elution from immobilized avidin supports (cleaving disulfide bond releases proteins)
  • Long spacer arm reduces steric hindrance for binding interactions
TRUSTED_SUSTAINABILITY

Specifications

Cleavable Yes
Description EZ-Link Sulfo-NHS-SS-Biotin, No-Weigh Format
Formulation Sulfo-NHS ester of biotin with an ethyl-1,3-dithioproprionate spacer
Molecular Weight (g/mol) 606.69
Membrane Permeable No
PEGylated No
Spacer Arm Length 24.3
Quantity 8 x 1 mg
Sufficient For 8 uses to label a total of 88mg of IgG using typical conditions
Content And Storage -20°C protected from moisture
Chemical Reactivity Amine
Solubility Water
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.