missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Sulfo NHS-SS Biotinylation Kit

Product Code. 10157723 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 Reactions
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10157723 10 Reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10157723 Supplier Thermo Scientific™ Supplier No. 21445

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biotinylate protein amines with this cleavable, mid-length (24.3A), water-soluble, NHS-ester biotinylation reagent containing a cleavable disulfide bond in the spacer arm.

Thermo Scientific™ EZ-Link Sulfo-NHS-SS-Biotin enables simple and efficient biotin labeling of antibodies, proteins and any other primary amine-containing biomolecules in solution.

Highlights:

  • Cleavable analog of Sulfo-NHS-LC-Biotin
  • Can remove biotin label by cleaving spacer disulfide bond with reducing agents, such as DTT
  • Ideal for affinity purification experiments that require efficient elution from immobilized avidin supports (cleaving disulfide bond releases proteins)
  • Long spacer arm reduces steric hindrance for binding interactions
TRUSTED_SUSTAINABILITY

Specifications

Description EZ-Link Sulfo-NHS-SS-Biotinylation Kit
Product Type Sulfo-NHS-SS Biotinylation Kit
Quantity 10 Reactions
Content And Storage Sufficient For: 10 labeling reactions, each with 1 to 10 mg of antibody
• EZ-Link Sulfo-NHS-SS-Biotin, 25 mg
• PBS Pack (makes 500 mL), 1 pack
• Zeba Spin Desalting Columns (7K MWCO), 5 mL, 10 columns
• HABA Solution, 1 mL
• Avidin, 10 mg

Store avidin at -20°C. Store biotin reagent desiccated at 4°C. Store remaining kit components at 4°C.
Labeling Scale 1 to 10 mg
Product Line EZ-Link
Solubility Water
Label or Dye Biotin
Labeling Target Antibodies, Proteins
compliance-icons
Product Identifier
  • EZ-Link Sulfo NHS-SS Biotinylation Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: disodium hydrogenorthophosphate sodium chloride

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.