missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ EZ-Link™ Micro Sulfo-NHS-SS-Biotinylation Kit
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ EZ-Link Sulfo-NHS-SS-Biotin enables simple and efficient biotin labeling of antibodies, proteins and any other primary amine-containing biomolecules in solution.
Highlights:
- Cleavable analog of Sulfo-NHS-LC-Biotin
- Can remove biotin label by cleaving spacer disulfide bond with reducing agents, such as DTT
- Ideal for affinity purification experiments that require efficient elution from immobilized avidin supports (cleaving disulfide bond releases proteins)
- Long spacer arm reduces steric hindrance for binding interactions
Specifications
Specifications
| Description | EZ-Link Micro Sulfo-NHS-SS-Biotinylation Kit |
| Product Type | Micro Sulfo-NHS-SS-Biotinylation Kit |
| Quantity | 8 reactions |
| Content And Storage | Sufficient For: 8 labeling reactions, each with 0.05–0.2 mg of antibody • EZ-Link Sulfo-NHS-SS-Biotin, 8 x 1 mg • PBS Pack (makes 500 mL), 1 pack • Zeba Spin Desalting Columns (7K MWCO), 2 mL, 10 columns Store biotin reagent at -20°C. Store remaining kit components at 4°C. |
| Labeling Scale | 100 μg |
| Product Line | EZ-Link |
| Solubility | Water |
| Label or Dye | Biotin |
| Labeling Target | Antibodies, Proteins |
Safety and Handling
Product Identifier
- EZ-Link Micro Sulfo-NHS-SS-Biotinylation Kit
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contain: Sodium phosphate dibasic, Sodium chloride
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?