missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Micro Sulfo-NHS-Biotinylation Kit

Product Code. 10687714 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
8 reactions
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10687714 8 reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10687714 Supplier Thermo Scientific™ Supplier No. 21925

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Biotinylate protein amines (e.g., lysines) with our shortest (13.5A), water-soluble, NHS-ester biotinylation reagent for antibody or cell-surface labeling.

Label antibodies, proteins and other molecules that have primary amines with Thermo Scientific™ EZ-Link Sulfo-NHS-Biotin, a short-chain, water-soluble biotinylation reagent.

Highlights:

  • Shorter spacer arm than Sulfo-NHS-LC-Biotin
  • Reacts with primary amines at pH 7-9 in non-amine-containing buffers such as borate, carbonate, or phosphate buffers
  • Negative charge of -SO3 keeps biotinylation localized at the cell surface so reagent does not pass through cell membrane
  • Water solubility eliminates the need for harmful organic solvents
U.S. Patents 5,892,628 and 5,872,261.
TRUSTED_SUSTAINABILITY

Specifications

Description EZ-Link Micro Sulfo-NHS-Biotinylation Kit
Product Type Micro Sulfo-NHS-Biotinylation Kit
Quantity 8 reactions
Content And Storage Sufficient For: 8 labeling reactions, each with 0.05–0.2 mg of antibody
• EZ-Link Sulfo-NHS-Biotin, 8 x 1 mg
• PBS Pack (makes 500 mL), 1 pack
• Zeba Spin Desalting Columns (7K MWCO), 2 mL, 10 columns

Store biotin reagent at -20°C. Store remaining kit components at 4°C.
Labeling Scale 100 μg
Product Line EZ-Link
Solubility Water
Label or Dye Biotin
Spacer Pegylated
Labeling Target Antibodies, Proteins
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.