missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ NHS-SS-Biotin

Product Code. 10608094 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 10608094

Brand: Thermo Scientific™ 21441

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biotinylate protein amines with this cleavable, mid-length (24.3A), NHS-ester biotinylation reagent containing a reducible disulfide bond in the membrane-permeable spacer arm.

Thermo Scientific™ EZ-Link NHS-SS-Biotin enables simple and efficient biotinylation of antibodies, proteins and other primary amine-containing molecules, as well as intracellular labeling.

Highlights:

  • Membrane-permeable analog of Sulfo-NHS-SS-Biotin
  • Dissolve in organic solvent (DMSO or DMF) for use
  • Dissolved reagent may be diluted into aqueous reaction buffer
  • Long spacer arm reduces steric hindrance for binding interactions
  • Disulfide bond is cleavable with DTT or other reducing agents
TRUSTED_SUSTAINABILITY

Specifications

Product Type NHS-SS-Biotin
Quantity 50 mg
Content And Storage Store at -20°C. Shipped at ambient temperature.
Product Line EZ-Link
Cell Permeability Cell-Permeant
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Label or Dye Biotin
Spacer Mid-length, Cleavable
Label Type Biotin & Analogs
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.