missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ NHS-PEG4-Biotin and Labeling Kits

Product Code. 11891195 Shop All Thermo Scientific Products
Click to view available options
Quantity:
8 x 2 mg
Unit Size:
Pack of 8
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11891195

Brand: Thermo Scientific™ 21329

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Biotinylate protein amines and maximize solubility of proteins with this long (29.0A) NHS-ester biotinylation reagent containing a pegylated, water-soluble spacer arm.

Thermo Scientific™ EZ-Link NHS-PEG4-Biotin is a pegylated, water-soluble reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.

Highlights:

  • Water-soluble polyethylene glycol (PEG) spacer arm increases solubility of labeled protein
  • Prevents aggregation of antibodies better than Sulfo-NHS-LC-Biotin
  • Labels at N-terminus or lysine residues in less than 1 hour
  • Thermo Scientific No-Weigh Packaging (8 x 2mg) saves time - preserves reagent activity and prevents cross-contamination between use
TRUSTED_SUSTAINABILITY

Specifications

Cleavable No
Description EZ-Link NHS-PEG4-Biotin, No-Weigh Format
Formulation NHS ester of biotin with a tetraethyleneglycol spacer
Molecular Weight (g/mol) 588.67
Membrane Permeable No
PEGylated Yes
Spacer Arm Length 29
Quantity 8 x 2 mg
Sufficient For 8 uses to label a total of 181mg of IgG using typical conditions
Content And Storage Store in original bottle at 4°C with desiccant
Chemical Reactivity Amine
Solubility Water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.