Learn More
EYS/RP25 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NBP1-90038
This item is not returnable.
View return policy
Description
EYS/RP25 Polyclonal specifically detects EYS/RP25 in Human, Zebrafish samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Immunohistochemistry, Immunocytochemistry | |
| Rabbit | |
| RUO |
| Polyclonal | |
| 0.1 mL | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |