missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Eye-wear Case with Belt Loop

Product Code. 11911661 Shop All Bürkle Products
Change view
Click to view available options
For Use With (Equipment):
For All Spectacles
For All Spectacles and Panoramic Safety Goggles
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11911661 For All Spectacles and Panoramic Safety Goggles Each
11901661 For All Spectacles Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11911661 Supplier Buerkle™ Supplier No. 25021009
  • ONLINE EXCLUSIVE PRICE
    £22.85
    £22.41 / Pack of 1
    Save £0.44
    1% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Buerkle™ Eye-wear Case with Belt Loop is extremely robust with reinforced zipper and is suitable for all spectacles.

  • Extremely robust eye-wear case
  • Includes reinforced zipper and belt loop
  • Suitable for all spectacles
  • Not for safety goggles
TRUSTED_SUSTAINABILITY

Specifications

Description Eye-wear case
Color Black
Product Type Eyewear Case
For Use With (Equipment) For All Spectacles and Panoramic Safety Goggles
Includes Belt loop
No. per Pack 1 Ea.
Disposable Reusable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.