missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Eye-wear Case with Belt Loop
Shop All Bürkle Products
Click to view available options
For Use With (Equipment):
For All Spectacles
For All Spectacles and Panoramic Safety Goggles
Unit Size:
Each
Description
- Extremely robust eye-wear case
- Includes reinforced zipper and belt loop
- Suitable for all spectacles
- Not for safety goggles
Specifications
Specifications
| Description | Eye-wear case |
| Color | Black |
| Product Type | Eyewear Case |
| For Use With (Equipment) | For All Spectacles |
| Includes | Belt loop |
| No. per Pack | 1 Ea. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction