Learn More
SYBR™ GreenER™ EXPRESS SYBR™ GreenER™ qPCR Supermix, universal
Kits are compatible with fast-cycling instruments for SYBR-based qPCR experiments.
Brand: Invitrogen™ 1178401K
981.00 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : 50
Additional Details : Weight : 0.50000kg
Description
EXPRESS SYBR GreenER qPCR SuperMix Universal comes with separate tubes of ROX reference dye.
- Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
- SYBR GreenER dye
- UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Specifications
Contains 5 × 5 ml EXPRESS SYBR™ GreenER™ qPCR SuperMix Universal (2X) and 5 × 500 μl ROX Reference Dye. Sufficient reagents are provided for 2500 reactions of 20 μL each. Store all components at -20°C for long-term storage. EXPRESS qPCR SuperMixes may be stored at 4–8°C for up to one month. |
|
Tube | |
qPCR | |
Fast or Standard | |
SYBR™ GreenER™ | |
2500 Reactions of 20 μL Each | |
2X | |
DNA (Genomic), cDNA |
SYBR | |
High | |
Taq DNA Polymerase | |
7500 Fast System, 7500 System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, BioRad CFX96™, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, BioRad MyiQ, Corbett RotorGene, Cepheid SmartCycler, MJ Opticon, MJ Chromo4, BioRad iCycler iQ | |
Real Time PCR SYBR Master Mix | |
Gene Expression | |
1000 Reactions | |
5 x 5 mL |
For Research Use Only. Not for use in diagnostic procedures.