missing translation for 'onlineSavingsMsg'
Learn More

SYBR™ GreenER™ EXPRESS SYBR™ GreenER™ qPCR Supermix, universal

Product Code. p-7083438
Click to view available options
No. of Reactions:
1000 Reactions
200 Reactions
Unit Size:
1000 reactions
Each
This item is not returnable. View return policy

Product Code. 10721734

Brand: SYBR™ GreenER™ 11784200

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Kits are compatible with fast-cycling instruments for SYBR-based qPCR experiments.

EXPRESS SYBR GreenER qPCR SuperMix Universal comes with separate tubes of ROX reference dye.

  • Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
  • SYBR GreenER dye
  • UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Contains 5 ml EXPRESS SYBR™ GreenER™ qPCR SuperMix Universal (2X) and 500 μl ROX Reference Dye. Sufficient reagents are provided for 500 reactions of 20 μL each.

Store all components at -20°C for long-term storage. EXPRESS qPCR SuperMixes may be stored at 4–8°C for up to one month.
Detection Method SYBR
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) 7500 Fast System, 7500 System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, BioRad CFX96™, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, BioRad MyiQ, Corbett RotorGene, Cepheid SmartCycler, MJ Opticon, MJ Chromo4, BioRad iCycler iQ
Product Line SYBR GreenER
Product Type Real Time PCR SYBR Master Mix
Quantity 200 reactions
Sufficient For 500 Reactions of 20 μL Each
For Use With (Application) Gene Expression
No. of Reactions 200 Reactions
Sample Type DNA (Genomic), cDNA
Volume 5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.