missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Exosome Precipitation Solution for enrichment of exosomes from serum or plasma
Click to view available options
Quantity:
12 mL Precipitation solution (with 30 mL RNase-free water)
2 mL Precipitation solution (with 13 mL RNase-free water)
60 mL Precipitation Solution (with 60 mL RNase-free water)
Unit Size:
12mL
2mL
60mL
Description
Exosomes are small vesicles (30–150 nm) secreted by various cell types and are found in many body fluids, such as blood, cerebrospinal fluid, urine, saliva, and breast milk. Their RNA and protein content can be a valuable source of biomarkers. By precipitating the exosomes from large sample volumes, those biomarkers can be enriched and the background of free circulating nucleic acids is significantly reduced.
- Most efficient enrichment of exosomes for any downstream application
- Simple and fast precipitation of exosomes – avoid tedious ultra centrifugation
- Flexibility in sample amount – procedure can be scaled up or down depending on sample volume
- Supplied with RNase-free water to isolate RNA from resuspended exomes
- Total preparation time: 45 min / 6 preps
Specifications
Specifications
| Color | Colorless |
| Includes | Exome Precipitation Solution; RNase-free Water |
| Odor | Odorless |
| Particle Size | 350 to 150 nm |
| Content And Storage | Room Temperature (18°C to 25°C) |
| Donor Source | Serum or Plasma Samples |
| Form | Liquid |
| For Use With (Application) | Enrichment of exosomes from blood serum and plasma samples |
| Sufficient For | 0.1 to 1 mL |
| Quantity | 60 mL Precipitation Solution (with 60 mL RNase-free water) |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion