missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Exosome Precipitation Solution for Serum/Plasma

Fast and efficient enrichment of exosomes from blood serum and plasma samples for the most efficient total RNA isolation or other biological studies. Includes RNase-free water.

Brand:  Macherey-Nagel™ 740398.12

Code : NEW

Additional Details : Weight : 0.00120kg

 View more versions of this product

Product Code. 15684454

  • £249.00 / 12mL
Estimated Shipment: 02-07-2024
to see stock.
Add to basket

Description

Description

Exosomes are small vesicles (30–150 nm) secreted by various cell types and are found in many body fluids, such as blood, cerebrospinal fluid, urine, saliva, and breast milk. Their RNA and protein content can be a valuable source of biomarkers. By precipitating the exosomes from large sample volumes, those biomarkers can be enriched and the background of free circulating nucleic acids is significantly reduced.

  • Most efficient enrichment of exosomes for any downstream application
  • Simple and fast precipitation of exosomes – avoid tedious ultra centrifugation
  • Flexibility in sample amount – procedure can be scaled up or down depending on sample volume
  • Supplied with RNase-free water to isolate RNA from resuspended exomes
  • Total preparation time: 45 min / 6 preps
Specifications

Specifications

Colorless
Odorless
Room Temperature (18°C to 25°C)
Liquid
0.1 to 1 mL Serum/Plasma Samples
Polyethylene glycol with chemicals; Water
Exome Precipitation Solution; RNase-free Water
350 to 150 nm
Serum or Plasma Samples
Enrichment of exosomes from blood serum and plasma samples
12 mL Precipitation solution (with 30 mL RNase-free water)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers