missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exosome component 7 Rabbit anti-Human, Mouse, Clone: 6E5N8, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Exosome component 7 |
|---|---|
| Clone | 6E5N8 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18379875
|
Novus Biologicals
NBP3-15270-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18309485
|
Novus Biologicals
NBP3-15270-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Exosome component 7 Monoclonal antibody specifically detects Exosome component 7 in Human, Mouse samples. It is validated for Western BlotSpecifications
| Exosome component 7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 23016 | |
| IgG | |
| Affinity purified |
| 6E5N8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EAP1, exosome complex exonuclease RRP42, exosome component 7Rrp42p, KIAA0116hRrp42p, p8FLJ26543, RRP42Ribosomal RNA-processing protein 42 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Exosome component 7 (Q15024). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title