missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exosome component 7 Rabbit anti-Human, Mouse, Clone: 6E5N8, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-15270-100UL
This item is not returnable.
View return policy
Description
Exosome component 7 Monoclonal antibody specifically detects Exosome component 7 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Exosome component 7 | |
| Monoclonal | |
| Unconjugated | |
| EAP1, exosome complex exonuclease RRP42, exosome component 7Rrp42p, KIAA0116hRrp42p, p8FLJ26543, RRP42Ribosomal RNA-processing protein 42 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Exosome component 7 (Q15024). | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 23016 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 6E5N8 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction