missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exosome component 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00
Specifications
| Antigen | Exosome component 1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228736
|
Novus Biologicals
NBP3-35122-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230892
|
Novus Biologicals
NBP3-35122-100ul |
100 μL |
N/A
|
N/A | |||||
Description
Exosome component 1 Polyclonal antibody specifically detects Exosome component 1 in Human samples. It is validated for ELISA,Western BlotSpecifications
| Exosome component 1 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| PBS (pH 7.3), 50% glycerol | |
| 51013 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CGI-108,3'-5' exoribonuclease CSL4 homolog, CSL4 exosomal core protein homolog, Csl4p, CSL4p13, exosomal core protein CSL4, exosome component 1hCsl4p, homolog of yeast exosomal core protein CSL4, SKI4, Ski4p | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-195 of human Exosome component 1 (NP_057130.1).,, Sequence:, MAPPVRYCIPGERLCNLEEGSPGSGTYTRHGYIFSSLAGCLMKSSENGALPVVSVVRETESQLLPDVGAIVTCKVSSINSRFAKVHILYVGSMPLKNSFRGTIRKEDVRATEKDKVEIYKSFRPGDIVLAKVISLGDAQSNYLLTTAENELGVVVAHSESGIQMVPISWCEMQCPKTHTKEFRKVARVQPEFLQT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title