missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exosome component 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35122-100ul
This item is not returnable.
View return policy
Description
Exosome component 1 Polyclonal antibody specifically detects Exosome component 1 in Human samples. It is validated for ELISA,Western Blot
Specifications
| Exosome component 1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| CGI-108,3'-5' exoribonuclease CSL4 homolog, CSL4 exosomal core protein homolog, Csl4p, CSL4p13, exosomal core protein CSL4, exosome component 1hCsl4p, homolog of yeast exosomal core protein CSL4, SKI4, Ski4p | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-195 of human Exosome component 1 (NP_057130.1).,, Sequence:, MAPPVRYCIPGERLCNLEEGSPGSGTYTRHGYIFSSLAGCLMKSSENGALPVVSVVRETESQLLPDVGAIVTCKVSSINSRFAKVHILYVGSMPLKNSFRGTIRKEDVRATEKDKVEIYKSFRPGDIVLAKVISLGDAQSNYLLTTAENELGVVVAHSESGIQMVPISWCEMQCPKTHTKEFRKVARVQPEFLQT | |
| 100 μL | |
| Immunology | |
| 51013 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction