missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOSC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-91874-25ul
This item is not returnable.
View return policy
Description
EXOSC3 Polyclonal antibody specifically detects EXOSC3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| EXOSC3 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50-1:200 | |
| bA3J10.7, CGI-102, exosome complex exonuclease RRP40, exosome component 3MGC15120, exosome component Rrp40, hRrp-40, hRrp40p, p10Rrp40p, Ribosomal RNA-processing protein 40, RP11-3J10.8, RRP40MGC723 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: VYWVDSQQKRYVPVKGDHVIGIVTAKSGDIFKVDVGGSEPASLSYLSFEGATKRNRPNVQVGDLIYGQFVVANKD | |
| 25 μL | |
| DNA replication Transcription Translation and Splicing | |
| 51010 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction