missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EXOSC10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | EXOSC10 |
|---|---|
| Concentration | 0.24 mg/mL |
| Applications | Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16896454
|
Proteintech
16731-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16886454
|
Proteintech
16731-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human DNA-binding protein (dbpA) is a member of a Y-box binding protein family containing a cold shock domain. The increased expression of Y box binding proteins in somatic cells is associated with cell proliferation and transformation.Specifications
| EXOSC10 | |
| Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P56960, Q01780 | |
| 50912, 5394 | |
| EXOSC10 Fusion Protein Ag10189 | |
| Primary | |
| -20°C | |
| EXOSC10 |
| 0.24 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Autoantigen PM/Scl 2, EXOSC10, exosome component 10, p2, p3, p4, PM Scl, PM/Scl 100, PMSCL, PMSCL2, RRP6, Rrp6p | |
| Exosc10 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title