Learn More
EXOSC10 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16731-1-AP-150UL
 View more versions of this product
Product Code. 16886454
  • £349.00 / 150µL
Description
Human DNA-binding protein (dbpA) is a member of a Y-box binding protein family containing a cold shock domain. The increased expression of Y box binding proteins in somatic cells is associated with cell proliferation and transformation.
Specifications
Specifications
EXOSC10
Polyclonal
Unconjugated
EXOSC10
Autoantigen PM/Scl 2, EXOSC10, exosome component 10, p2, p3, p4, PM Scl, PM/Scl 100, PMSCL, PMSCL2, RRP6, Rrp6p
Rabbit
Antigen Affinity Chromatography
RUO
50912, 5394
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P56960, Q01780
Exosc10
EXOSC10 Fusion Protein Ag10189
150 μL
Primary
Human, Mouse