missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXOC3 Recombinant Rabbit Monoclonal Antibody (23GB5345), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00
Specifications
| Antigen | EXOC3 |
|---|---|
| Clone | 23GB5345 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
Description
EXOC3 Recombinant Monoclonal antibody specifically detects EXOC3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| EXOC3 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O60645, Q62825, Q6KAR6 | |
| 11336, 211446, 252881 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5345 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2810050O03Rik; E430013E20Rik; EXOC3; EXOC3L; EXOC3L1; Exocyst complex component 3; exocyst complex component 3 like 1; exocyst complex component 3-like 1; exocyst complex component 3-like protein; Exocyst complex component Sec6; I79_024581; KIAA0895L; KIAA0895-like ortholog; LIM/homeobox protein Lhx2 pseudogene; protein Jiangli; RGD1311132; rSec6; Sec 6 homolog; Sec6; Sec6 protein; SEC6L1; SEC6-like 1; Sec6p | |
| A synthesized peptide derived from human EXOC3 (680-700AA). | |
| Primary | |
| -20°C | |
| EXOC3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title