missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXOC3 Recombinant Rabbit Monoclonal Antibody (23GB5345), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552840
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity.
Specifications
| EXOC3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| EXOC3 | |
| 2810050O03Rik; E430013E20Rik; EXOC3; EXOC3L; EXOC3L1; Exocyst complex component 3; exocyst complex component 3 like 1; exocyst complex component 3-like 1; exocyst complex component 3-like protein; Exocyst complex component Sec6; I79_024581; KIAA0895L; KIAA0895-like ortholog; LIM/homeobox protein Lhx2 pseudogene; protein Jiangli; RGD1311132; rSec6; Sec 6 homolog; Sec6; Sec6 protein; SEC6L1; SEC6-like 1; Sec6p | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11336, 211446, 252881 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5345 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60645, Q62825, Q6KAR6 | |
| EXOC3 | |
| A synthesized peptide derived from human EXOC3 (680-700AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction