missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exendin-4 Antibody (20) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£175.00 - £349.00
Specifications
| Antigen | Exendin-4 |
|---|---|
| Clone | 20 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18143945
|
Novus Biologicals
NBP2-23577-0.025ML |
0.025 mL |
£175.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18135093
|
Novus Biologicals
NBP2-23577-0.1ML |
0.1 mL |
£349.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Exendin-4 Monoclonal specifically detects Exendin-4 in Reptile samples. It is validated for ELISA.Specifications
| Exendin-4 | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
| Synthetic exendin-4 adsorbed onto aluminum hydroxide gel | |
| Primary | |
| Store at 4C. Do not freeze. |
| 20 | |
| ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| Human | |
| Exendin4 | |
| IgG1 κ | |
| Protein A or G purified | |
| Specific for exendin-4. No cross-reactivity with GLP-1, GLP-2 (human) or glucagon coated on ELISA wells |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title