missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exendin-4 Antibody (20) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23577-0.025ML
This item is not returnable.
View return policy
Description
Exendin-4 Monoclonal specifically detects Exendin-4 in Reptile samples. It is validated for ELISA.
Specifications
| Exendin-4 | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:100-1:2000 | |
| Exendin4 | |
| Synthetic exendin-4 adsorbed onto aluminum hydroxide gel | |
| 0.025 mL | |
| Specific for exendin-4. No cross-reactivity with GLP-1, GLP-2 (human) or glucagon coated on ELISA wells | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
| ELISA | |
| 20 | |
| Unconjugated | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction