missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Evodiamine

Product Code. 15465069
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465069 250 mg 250mg
15455069 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465069 Supplier Thermo Scientific Chemicals Supplier No. J62771.MD
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 518-17-2 | C19H17N3O, C19H17N3O | 303.37 g/mol

A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 518-17-2
Molecular Formula C19H17N3O

Specifications

Quantity 250 mg
Description Off-white to pale yellow
Content And Storage -30°C to -10°C
Format Solid
For Use With (Application) A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells
Formula Weight 303.40
Product Type Evodiamine
Form Solid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.