missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ EVA Foam Ice Pans and Buckets with Lids

Product Code. 16574251 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
1 L
2.5 L
4 L
9 L
Color:
Blue
Green
Purple
Includes:
Bucket with Lid
Pan with Lid
Unit Size:
Each

Product Code. 16574251

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Includes

removable lid.

Fisherbrand™EVA Foam Ice Pans and Buckets with Lids provide excellent insulation for any benchtop cryogenic application, without the mess!

  • Sweat-proof ethylene vinyl acetate will not leak
  • Compatible with ice, dry ice, liquid nitrogen, alcohol, and saline solutions
  • Bases and lids feature glove-friendly handles
  • Cleanup is quick and easy with molded pour spouts

Specifications

For Use With (Equipment) Ice, dry ice, liquid nitrogen, alcohol, and saline solutions
Includes Bucket with Lid
Description Bucket with Lid
Material EVA
Type Cryo-Safe Cold Box
Quantity 1
Color Purple
Capacity (Metric) 4 L
Autoclavable Not Autoclavable
Temperature Control No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.