missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ EV Precipitation Solution (Blood)

Product Code. 18624330 Shop All Bio Techne Products
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 18624330

Brand: Novus Biologicals™ NBP311765

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Product Type EV isolation, exosome isolation, extracellular vesicle isolation
Quantity 5 mL
Sufficient For A 5mL solution is equivalent to 180 reactions (plasma) and 80 reactions (serum).
Content And Storage Store at room temperature.
Sample Type Plasma and serum
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.