missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ PD 450 Meter

Product Code. 15307073
Change view
Click to view available options
Product Type:
Meter with Electrodes
Multiparameter Meter
Unit Size:
Each
Product Code. Product Type unitSize
15307073 Multiparameter Meter Each
15327073 Meter with Electrodes Each
2 options
This item is not returnable. View return policy

Product Code. 15307073

Brand: Thermo Scientific™ ECPDWP45000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Collecting multi-parameter data is a breeze using Eutech™ PD 450 Meter.

The Eutech™ PD450 Meter is a multi-parameter meter that measures pH, mV, dissolved oxygen, and temperature. Affordable optical RDO technology reduces maintenance time and costs. The maintenance free optical probe with no electrolyte eliminates the hassle of replacing delicate membranes and salt build up from refillling solutions. You can also easily switch between pH, mV, % saturation and mg/L (ppm) dissolved oxygen – with the press of a button.

Features:

  • pH / mV / Dissolved Oxygen / Temperature
  • Durable waterproof and dustproof design with IP67 rating
  • Large, backlit graphic display provides measurement with temperature
  • Calibrate up to 5 points automatically with USA or NIST buffer groups (up to 6 points with DIN). New manual option allows calibration with up to 5 custom buffer standards
  • Auto buffer recognition – Automatically identifies the correct pH buffer for rapid calibration
  • Calibration due alarm – Set a visible reminder when it is time to recalibrate the meter
  • Selectable ºC or ºF – Choose units to best fit your application
  • Ready indicator, manual or automatic measurement hold feature and optional automatic shutoff function
  • Store up to 500 data sets – Easy USB or RS232 output with optional data cables
  • Built-in real-time clock stamps stored data and calibration data with time and date – Meets standards for GLP (Good Laboratory Practices)
TRUSTED_SUSTAINABILITY

Specifications

AC Adapter Optional
Outputs USB or RS232
Accuracy (DO) ±2 %; ±0.1 ppm or mg/L up to 8; ±0.2 ppm or mg/L above 8
Accuracy (mV) ±0.2 mV +2 LSD or +0.5 % of reading; whichever is greater
Accuracy (pH) ±0.01
Battery Life 100+ hr. (without backlight)
Battery Type 2 AA (included)
Calibration mV: ±200 mV, DO: Zero and saturation, Temp: Offset in 0.1 increments, ±10 °C or &
Calibration Points (pH) Up to 6 points: USA, NIST, DIN or custom
Compensation Pressure & Salinity Compensation: manual
Description PD 450 Meter Only: Meter, two AA batteries
Display Type Backlit Custom LCD
IP Rating IP67
Measurement Modes pH, mV, Dissolved Oxygen, Temperature
Range (mV) ±2000 mV
Range (pH) -2 to 16
Resolution (pH) 0.01
Temperature Range (Metric) -10.0°C to +110.0°C
Includes Time and date stamp on calibration and stored data
Memory Stores up to 500 data sets
Product Type Multiparameter Meter
Range (Polarographic DO) 0 to 200 %; 0 to 20 ppm/mg/L
Resolution (mV) 0.1 mV ±999.9 mV; 1 mV beyond
Resolution (Polarographic DO) 0.10 %; 0.1 ppm/mg/L
Temperature Accuracy ±0.5°C, ±0.9°F
Temperature Compensation Automatic or manual from -10 to 110 °C
Temperature Range (English) 14.0°F to 230.0°F
Temperature Resolution 0.1 ºC/ºF
Dimensions (L x W x H) Meter: 7 7/8in x 3 1/8in x 2 1/4in (20 x 8.3 x 5.7 cm), Boxed: 9 1/8in x 9 1/8in x 2 3/4in (23 x 23 x 7 cm)
Weight (English) Meter: 1 lb , Boxed: 1.4 lb
Weight (Metric) Meter: 0.5 kg, Boxed: 0.7 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.