Learn More
Thermo Scientific™ Eutech™ PCTestr 35 Multi-Parameter Pocket Tester
Description
No need to change sensor every time you switch parameters – The Eutech™ PCTestr 35 Multi-Parameter Pocket Tester measures three parameters (measures pH, Conductivity and Temperature). For general applications: Ideal for quick and accurate pH checks in pools and spas, aquariums and hydroponics operations, or anywhere frequent pH testing is required. PCSTestr 35 meets requirements for many standard environmental and ASTM test methods. For industrial applications: Use for cooling towers, food processing water testing and process/wastewater testing in metal finishing, photo development, printing and chemical industries. For educational applications: Useful in schools, many laboratory applications and ecology studies.
Product Features
- Full pH range measurement at up to 2-decimal places.
- Low, medium and high Conductivity ranges – tester measures pure water!
- Up to 5-point pH calibration and 3-point Conductivity calibration.
- Adjustable Temperature Coefficient (0.0 to 0.0%)
- Temperature sensor housed within unique dome-shaped casing for faster ATC response and faster measurements.
- ATC/ MTC option
- Scroll-menu with four buttons control for stress-free navigation
- Longest-lasting tester in the market! No tools required to replace sensor.
- One-press Parameter Toggle: Switching parameters is just a press of the button.
- Certified IP67 Waterproof – Lightweight tester even floats for easy retrieval.
Specifications
Specifications
| Accuracy (Conductivity) | ± 1% full scale |
| Accuracy (pH) | ±0.01 |
| Accuracy (Salinity) | 1% full scale |
| Accuracy (TDS) | 1% full scale |
| Battery Life | 150+ hours |
| Battery Type | 4 X 1.5 V A76 Micro Alkaline |
| Calibration Points | TDS: Up to 3 Points (Manual), Salinity: Up to 3 (Manual) |
| Calibration Points (Conductivity) | 3 Auto, 3 Manual |
| Calibration Points (pH) | Up to 5 (Auto), Buffer Options: USA, NIST |
| Description | Waterproof PCTestr 35 pH/Conductivity/Temp Tester with carrying kit |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.