missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ Ion 700 Meter

Product Code. 11523855
Change view
Click to view available options
Measurement Modes:
pH, Redox, Ion
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Modes unitSize
11523855 pH, Redox, Ion Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11523855 Supplier Thermo Scientific™ Supplier No. 01X541609

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Quick and easy measurement with this ion-selective meter. The Ion 700 measures and records up to 100 data points with 0.01 resolution.

Meter accepts pH, Redox or Ion selective electrodes– like having three meters in one. It has a large, easy-to-read display with integral electrode holder. Direct ion concentration measurement– use with many different Ion-selective electrodes. Up to 5-point calibration with auto-buffer recognition and the non-volatile memory holds up to 100 data points.

Specifications

pH

  • Range: -2.00 to 16.00 pH
  • Resolution: 0.01 pH
  • Accuracy: ±0.01 pH
  • Cal. Points: Up to 5
  • Buffer Sets: USA (1.68, 4.01, 7.00, 10.01, 12.45), NIST (1.68, 4.01, 6.86, 9.18, 12.45)

ORP

  • Range: ±2000mV
  • Rel.mV Range: ±2000mV
  • Resolution: 0.1mV (±199.9mV) / 1mV (beyond)
  • Accuracy: ±0.2mV (±199.9mV) / 2mV (beyond)

Ion

  • Concentration: 0.01 to 2000 ppm (±2000mV)
  • Resolution (ppm): 0.01 / 0.1 / 1 ppm
  • Accuracy: ±0.5 % full scale (monovalent), ±1 % full scale (divalent)
  • Cal. Points: 2 to 5

Temperature

  • Range: 0.0 to 100.0 °C / 32.0 to 212.0 °F
  • Resolution: 0.1 °C / 0.1 °F
  • Accuracy: ±0.3 °C / ±0.5 °F (0 to 70 °C)
  • Compensation: ATC /mTC (0 to 100 °C) (pH only)
  • Calibration: Offset in 0.1 ° increments; offset range: ±5.0 °C / ±9.0 °F
  • Power Requirements: 9 V DC adapter, 1.3 A (100/240 VAC, SmPS)
TRUSTED_SUSTAINABILITY

Specifications

Description pH, Redox and Ion selective meter
Measurement Modes pH, Redox, Ion
Resolution (pH) 0.01
Temperature Range (Metric) 0°C to 100°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.