Learn More
Etoposide, Thermo Scientific™
Topoisomerase inhibitor and inducer of apoptosis
£62.65 - £157.65
Identificadores de productos químicos
| CAS | 33419-42-0 |
|---|---|
| Molecular Formula | C29H32O13 |
| Molecular Weight (g/mol) | 588.56 |
| MDL Number | MFCD00869325,MFCD00869325 |
| InChI Key | VJJPUSNTGOMMGY-MRVIYFEKSA-N |
| Synonym | vjjpusntgommgy-nzlmilqcsa |
| PubChem CID | 50936917 |
| IUPAC Name | (10R,11R,15R,16S)-16-{[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one |
| SMILES | COC1=CC(=CC(OC)=C1O)[C@H]1[C@@H]2[C@H](COC2=O)[C@H](O[C@@H]2O[C@@H]3CO[C@@H](C)O[C@H]3[C@H](O)[C@H]2O)C2=CC3=C(OCO3)C=C12 |
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|
15495819
|
Thermo Scientific Alfa Aesar
J63651.MB |
25mg |
£69.70 £62.65 / 25mg Save £7.05 10% Off |
In Stock
Log in to see stock.
|
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15405829
|
Thermo Scientific Alfa Aesar
J63651.MC |
100mg |
£175.00 £157.65 / 100mg Save £17.35 9% Off |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsEtoposide is used as a inhibits DNA synthesis and is very active against cells in the late S and G2 phases of the cell cycle. Etoposide was reported to be used to serum starve Saos-2 M5(p53-/-) cells which were transfected by electroporation with indicated expression plasmids for flag-E2F1 and myc-EBNA3C for selection of colony forming units, thus studying the blocking effect of EBV Nuclear Antigen 3C on E2F1 Mediated Apoptosis Induced by the DNA Damage Response.
Solubility
Insoluble in water.
Notes
Store solutions in DMSO at 4°C. For long term storage prepare aliquots and store at -20°C.
Identificadores de productos químicos
| 33419-42-0 | |
| 588.56 | |
| VJJPUSNTGOMMGY-MRVIYFEKSA-N | |
| 50936917 | |
| COC1=CC(=CC(OC)=C1O)[C@H]1[C@@H]2[C@H](COC2=O)[C@H](O[C@@H]2O[C@@H]3CO[C@@H](C)O[C@H]3[C@H](O)[C@H]2O)C2=CC3=C(OCO3)C=C12 |
| C29H32O13 | |
| MFCD00869325,MFCD00869325 | |
| vjjpusntgommgy-nzlmilqcsa | |
| (10R,11R,15R,16S)-16-{[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one |
Especificaciones
| 33419-42-0 | |
| MFCD00869325,MFCD00869325 | |
| vjjpusntgommgy-nzlmilqcsa | |
| VJJPUSNTGOMMGY-MRVIYFEKSA-N | |
| COC1=CC(=CC(OC)=C1O)[C@H]1[C@@H]2[C@H](COC2=O)[C@H](O[C@@H]2O[C@@H]3CO[C@@H](C)O[C@H]3[C@H](O)[C@H]2O)C2=CC3=C(OCO3)C=C12 | |
| 588.56 | |
| 588.56 |
| C29H32O13 | |
| 14,3886 | |
| Insoluble in water. | |
| −107° (c=1 in Methanol/Dichloromethane 1:9) | |
| (10R,11R,15R,16S)-16-{[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one | |
| 50936917 | |
| Etoposide |
Seguridad y manipulación
GHS H Statement
H350-H302
May cause cancer.
Harmful if swallowed.
P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H350
missing translation for 'einecsNumber' : 251-509-1
missing translation for 'rtecsNumber' : KC0190000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.