Learn More
Ethyl bromopyruvate, 80-85%, Thermo Scientific Chemicals
£17.66 - £175.00
Chemical Identifiers
CAS | 70-23-5 |
---|---|
Molecular Formula | C5H7BrO3 |
Molecular Weight (g/mol) | 195.01 |
MDL Number | MFCD00000204 |
InChI Key | VICYTAYPKBLQFB-UHFFFAOYSA-N |
Synonym | ethyl bromopyruvate, ethyl 3-bromopyruvate, propanoic acid, 3-bromo-2-oxo-, ethyl ester, bromopyruvic acid ethyl ester, ethyl bromo pyruvate, ethyl 3-bromo-2-oxopropionate, pyruvic acid, bromo-, ethyl ester, .beta.-bromopyruvic acid ethyl ester, 3-bromo-2-oxo-propionic acid ethyl ester, beta-bromopyruvic acid ethyl ester |
PubChem CID | 66144 |
IUPAC Name | ethyl 3-bromo-2-oxopropanoate |
SMILES | CCOC(=O)C(=O)CBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10500751
|
Thermo Scientific Acros
118150050 |
5 mL | Glass bottle | ||||||
10655152
|
Thermo Scientific Acros
118150250 |
25 mL | Glass bottle |
£54.60
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10697933
|
Thermo Scientific Acros
118151000 |
100 mL | Glass bottle |
£175.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
70-23-5 | |
195.01 | |
VICYTAYPKBLQFB-UHFFFAOYSA-N | |
66144 | |
CCOC(=O)C(=O)CBr |
C5H7BrO3 | |
MFCD00000204 | |
ethyl bromopyruvate, ethyl 3-bromopyruvate, propanoic acid, 3-bromo-2-oxo-, ethyl ester, bromopyruvic acid ethyl ester, ethyl bromo pyruvate, ethyl 3-bromo-2-oxopropionate, pyruvic acid, bromo-, ethyl ester, .beta.-bromopyruvic acid ethyl ester, 3-bromo-2-oxo-propionic acid ethyl ester, beta-bromopyruvic acid ethyl ester | |
ethyl 3-bromo-2-oxopropanoate |
Specifications
70-23-5 | |
1.5500g/mL | |
98°C | |
80% min. (GC) | |
BrCH2COCO2C2H5 | |
MFCD00000204 | |
1.55 | |
VICYTAYPKBLQFB-UHFFFAOYSA-N | |
ethyl 3-bromo-2-oxopropanoate | |
66144 | |
80-85% | |
Ethyl bromopyruvate |
Pink-Red to Yellow | |
100.0°C to 102.0°C (15.0 mmHg) | |
Authentic | |
C5H7BrO3 | |
1.4675 to 1.4765 | |
03, II, 409 | |
ethyl bromopyruvate, ethyl 3-bromopyruvate, propanoic acid, 3-bromo-2-oxo-, ethyl ester, bromopyruvic acid ethyl ester, ethyl bromo pyruvate, ethyl 3-bromo-2-oxopropionate, pyruvic acid, bromo-, ethyl ester, .beta.-bromopyruvic acid ethyl ester, 3-bromo-2-oxo-propionic acid ethyl ester, beta-bromopyruvic acid ethyl ester | |
CCOC(=O)C(=O)CBr | |
195.01 | |
195.01 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-729-6
- Ethyl bromopyruvate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only