missing translation for 'onlineSavingsMsg'
Learn More

Ethyl bromopyruvate, 80-85%

Product Code. 10655152 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10655152 25 mL Glass bottle 25mL
10500751 5 mL Glass bottle 5mL
10697933 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10655152 Supplier Thermo Scientific Acros Supplier No. 118150250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70-23-5 | C5H7BrO3 | 195.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-23-5
Molecular Formula C5H7BrO3
Molecular Weight (g/mol) 195.01
MDL Number MFCD00000204
InChI Key VICYTAYPKBLQFB-UHFFFAOYSA-N
Synonym ethyl bromopyruvate, ethyl 3-bromopyruvate, propanoic acid, 3-bromo-2-oxo-, ethyl ester, bromopyruvic acid ethyl ester, ethyl bromo pyruvate, ethyl 3-bromo-2-oxopropionate, pyruvic acid, bromo-, ethyl ester, .beta.-bromopyruvic acid ethyl ester, 3-bromo-2-oxo-propionic acid ethyl ester, beta-bromopyruvic acid ethyl ester
PubChem CID 66144
IUPAC Name ethyl 3-bromo-2-oxopropanoate
SMILES CCOC(=O)C(=O)CBr

Specifications

Color Pink-Red to Yellow
Density 1.5500g/mL
Boiling Point 100.0°C to 102.0°C (15.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 80% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2COCO2C2H5
Refractive Index 1.4675 to 1.4765
Quantity 25 mL
Beilstein 03, II, 409
Specific Gravity 1.55
Formula Weight 195.01
Percent Purity 80-85%
Physical Form Liquid
Chemical Name or Material Ethyl bromopyruvate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl bromopyruvate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.